Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sez6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291225.1 | NP_001278154.1 | 3 | 1058 | ||
NM_021286.4 | NP_067261.2 | 3 | 1058 | |||
GenBank mRNA | AB206791.1 | - | 3 | 994 | ||
AK032277.1 | - | 3 | 982 | |||
AK147705.1 | - | 3 | 977 | |||
AK165757.1 | - | 3 | 842 | |||
AK220513.1 | - | 2 | 1688 | |||
BC053011.1 | - | 3 | 954 | 5698186 | ||
BC055345.1 | - | 3 | 1058 | 6833344 | ||
D29763.1 | - | 3 | 885 | |||
D64009.1 | - | 3 | 858 | |||
D64010.1 | - | 3 | 858 |