Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sez6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291225.1 | NP_001278154.1 | 15 | 3079 | ||
NM_021286.4 | NP_067261.2 | 16 | 3118 | |||
GenBank mRNA | AB206791.1 | - | 16 | 3054 | ||
AK147705.1 | - | 13 | 3440 | |||
AK161576.1 | - | 11 | 1528 | |||
AK165757.1 | - | 14 | 3193 | |||
AK220513.1 | - | 15 | 3748 | |||
BC053011.1 | - | 16 | 3014 | 5698186 | ||
BC055345.1 | - | 14 | 3441 | 6833344 | ||
D29763.1 | - | 15 | 2906 | |||
D64009.1 | - | 16 | 2918 | |||
D64010.1 | - | 15 | 3131 |