Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfrp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016687.3 | NP_057896.1 | 5 | 1071 | ||
GenBank mRNA | AF117709.1 | - | 5 | 1086 | ||
AK009981.1 | - | 5 | 1062 | |||
AK080766.1 | - | 5 | 1072 | |||
BC034853.1 | - | 5 | 1030 | 1364770 |
Set Membership: |
![]() |