Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161845.2 | NP_001155317.2 | 12, 13 | 1783 | ||
NM_001161847.2 | NP_001155319.1 | 10, 11 | 1284 | |||
NM_001161848.2 | NP_001155320.1 | 10, 11 | 1077 | |||
NM_001161849.2 | NP_001155321.1 | 10, 11 | 1158 | |||
NM_001161850.2 | NP_001155322.1 | 10, 11 | 1265 | |||
NM_011361.3 | NP_035491.1 | 10, 11 | 1133 | |||
GenBank mRNA | AF139638.1 | - | 10, 11 | 1117 | ||
AF205855.1 | - | 10, 11 | 1114 | |||
AK086892.1 | - | 10, 11 | 1117 | |||
AK132234.1 | - | 10, 11 | 1205 | |||
AK159131.1 | - | 10, 11 | 1117 | |||
AK167364.1 | - | 10, 11 | 1114 | |||
BC002222.1 | - | 3, 4 | 311 | |||
BC005720.1 | - | 10, 11 | 1094 | 3594892 | ||
BC070401.1 | - | 12, 13 | 1783 | 5695396 |