Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itsn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198968.2 | NP_001185897.1 | 25 | 3321 | ||
NM_011365.4 | NP_035495.2 | 24 | 3240 | |||
GenBank mRNA | AF132479.1 | - | 23 | 2980 | ||
AF132480.1 | - | 23 | 2980 | |||
AK122480.1 | - | 19 | 2627 | |||
AK161743.1 | - | 13 | 1658 | |||
BC138262.1 | - | 25 | 3298 | 8861281 | ||
BC171950.1 | - | 25 | 3298 | 9054236 |