Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009167.3 | NP_033193.2 | 4, 5 | 373 | ||
GenBank mRNA | BC103611.1 | - | 4, 5 | 373 | 40046304 | |
BC103612.1 | - | 4, 5 | 373 | 40046306 | ||
BC105644.1 | - | 4, 5 | 373 | 40046301 | ||
BC105645.1 | - | 4, 5 | 373 | 40046303 | ||
U46854.1 | - | 4, 5 | 373 |