Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110350.1 | NP_001103820.1 | 9 | 1669 | ||
NM_001110351.1 | NP_001103821.1 | 10 | 1546 | |||
NM_011378.2 | NP_035508.2 | 10 | 1777 | |||
GenBank mRNA | AK032209.1 | - | 9 | 1537 | ||
AK136347.1 | - | 4 | 471 | |||
AK220292.1 | - | 9 | 1646 | |||
BC052716.1 | - | 9 | 1556 | 6837144 | ||
BC053385.1 | - | 9 | 1539 | |||
BC078454.1 | - | 9 | 1530 | |||
L36831.1 | - | 9 | 1460 | |||
L38620.2 | - | 8 | 1436 | |||
L38621.2 | - | 8 | 1436 | |||
U22394.1 | - | 10 | 1785 |