Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253804.1 | NP_001240733.1 | 18 | 2349 | ||
NM_009195.3 | NP_033221.1 | 18 | 2343 | |||
GenBank mRNA | AF047339.1 | - | 18 | 2267 | ||
AF121118.1 | - | 18 | 2334 | |||
AK149969.1 | - | 18 | 2332 | |||
AK151717.1 | - | 18 | 2304 | |||
AK152762.1 | - | 18 | 2321 | |||
AK153245.1 | - | 18 | 2321 | |||
AK159274.1 | - | 18 | 2331 | |||
AK159331.1 | - | 18 | 2326 | |||
AK159483.1 | - | 18 | 2299 | |||
AK159537.1 | - | 18 | 2326 | |||
AK160034.1 | - | 18 | 2292 | |||
AK160072.1 | - | 18 | 2326 | |||
AK164479.1 | - | 18 | 2308 | |||
AK165874.1 | - | 18 | 2321 | |||
AK166304.1 | - | 18 | 2302 | |||
AK167371.1 | - | 18 | 2340 | |||
AK167444.1 | - | 18 | 2340 | |||
AK169044.1 | - | 18 | 2281 | |||
BC066872.1 | - | 18 | 2273 | 30436525 |