Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc20a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011394.3 | NP_035524.2 | 5 | 1068 | ||
GenBank mRNA | AF196476.1 | - | 4 | 836 | ||
AK087507.1 | - | 5 | 893 | |||
AK157206.1 | - | 5 | 942 | |||
AK170985.1 | - | 5 | 1034 | |||
AK198610.1 | - | 3 | 232 | |||
AK216704.1 | - | 1 | 79 | |||
BC046510.1 | - | 5 | 991 | 4035182 | ||
BC094562.1 | - | 5 | 1448 |