Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slfn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011409.1 | NP_035539.1 | 2 | 619 | ||
GenBank mRNA | AF099974.1 | - | 2 | 619 | ||
AK133208.1 | - | 2 | 1971 | |||
AK213262.1 | - | 1 | 89 | |||
BC132627.1 | - | 2 | 557 | 40130904 | ||
BC138070.1 | - | 2 | 557 | 8861089 | ||
BC145134.1 | - | 3 | 724 | 9053682 |