Gene Symbol: Slit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015748.3 | NP_056563.2 | 17 | 2103 | ||
GenBank mRNA | AF144627.1 | - | 17 | 2128 | ||
AK122376.1 | - | 17 | 1869 | |||
BC057131.1 | - | 17 | 1909 | 6827560 | ||
BC062091.1 | - | 17 | 1909 | 6827560 |