Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eftud2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109995.1 | NP_001103465.1 | 12, 13 | 1137 | ||
NM_011431.3 | NP_035561.1 | 12, 13 | 1134 | |||
GenBank mRNA | AK052242.1 | - | 12, 13 | 1092 | ||
AK150158.1 | - | 12, 13 | 1106 | |||
AK153473.1 | - | 12, 13 | 1106 | |||
AK154878.1 | - | 12, 13 | 1116 | |||
AK165624.1 | - | 12, 13 | 1108 | |||
AK172875.1 | - | 12, 13 | 1088 | |||
AK202817.1 | - | 2, 3 | 134 | |||
AK212622.1 | - | 2, 3 | 149 | |||
BC052674.1 | - | 12, 13 | 1110 | 30061580 | ||
BC054778.1 | - | 12, 13 | 1100 | 6810127 | ||
U97079.1 | - | 12, 13 | 1076 |