Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005045.2 | - | 11 | 2404 | ||
AK077425.1 | - | 10 | 3428 | |||
AK133115.1 | - | 10 | 1045 | |||
AK146729.1 | - | 10 | 1045 | |||
AK148013.1 | - | 10 | 1073 | |||
BC002169.1 | - | 4 | 223 | |||
BC049128.1 | - | 10 | 3376 | |||
BC058513.1 | - | 3 | 550 | |||
BC094067.1 | - | 3 | 701 | |||
BC131994.1 | - | 10 | 937 | 40130271 | ||
BC132020.1 | - | 10 | 937 | 40130297 |