Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000884.2 | NP_000875.2 | 11 | 1314 | ||
XM_006713128.3 | XP_006713191.1 | 12 | 1694 | |||
XM_017006349.1 | XP_016861838.1 | 11 | 1571 | |||
XM_017006350.1 | XP_016861839.1 | 10 | 1499 | |||
GenBank mRNA | BC006124.1 | - | 11 | 1262 | 3545693 | |
BC012840.1 | - | 11 | 1255 | 3447994 | ||
BC015567.2 | - | 11 | 1253 | 4576285 | ||
J04208.1 | - | 11 | 1269 |