Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000884.2 | NP_000875.2 | 4 | 376 | ||
XM_006713128.3 | XP_006713191.1 | 4 | 684 | |||
GenBank mRNA | AK293397.1 | - | 337 | |||
BC006124.1 | - | 4 | 324 | 3545693 | ||
BC012840.1 | - | 4 | 317 | 3447994 | ||
BC015567.2 | - | 4 | 315 | 4576285 | ||
J04208.1 | - | 4 | 331 |