Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013672.2 | NP_038700.2 | 2 | 211 | ||
GenBank mRNA | AF022363.1 | - | 2 | 199 | ||
AF062566.1 | - | 2 | 147 | |||
CJ181598.1 | - | 2 | 218 | |||
EF200003.1 | - | 2 | 346 | |||
EF200004.1 | - | 2 | 145 | |||
EF200005.1 | - | 2 | 215 | |||
EF200006.1 | - | 2 | 251 | |||
EF200007.1 | - | 1 | 607 | |||
EF200008.1 | - | 1 | 120 | |||
EF200009.1 | - | 1 | 198 | |||
S79832.1 | - | 2 | 202 |