Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021287.1 | NP_067262.1 | 37 | 7543 | ||
GenBank mRNA | AK147671.1 | - | 37 | 7559 | ||
AK172931.1 | - | 27 | 5852 | |||
BC033305.1 | - | 3 | 342 | |||
BC048851.1 | - | 15 | 2361 | |||
BC079628.1 | - | 15 | 2495 | |||
BC079860.1 | - | 37 | 7543 | 6834745 |