Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021287.1 | NP_067262.1 | 21 | 4937 | ||
GenBank mRNA | AK147671.1 | - | 21 | 4953 | ||
AK172931.1 | - | 11 | 3246 | |||
BC079860.1 | - | 21 | 4937 | 6834745 |