Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sprr2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011468.4 | NP_035598.2 | 2 | 2639 | ||
GenBank mRNA | AK147183.1 | - | 2 | 2555 | ||
AK168529.1 | - | 2 | 2552 |
Gene Symbol: Sprr2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164787.1 | NP_001158259.1 | 2639 |