Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sprr2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011477.3 | NP_035607.2 | 2 | 323 | ||
GenBank mRNA | BC116866.1 | - | 2 | 309 | ||
BC116868.1 | - | 2 | 309 |