Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: INHBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002192.2 | NP_002183.1 | 3 | 1209 | ||
XM_017012174.1 | XP_016867663.1 | 5 | 3101 | |||
XM_017012175.1 | XP_016867664.1 | 3 | 2785 | |||
XM_017012176.1 | XP_016867665.1 | 5 | 2969 | |||
GenBank mRNA | AF218018.1 | - | 3 | 1168 | ||
AK222742.1 | - | 2 | 1038 | |||
AK290584.1 | - | 3 | 1172 | |||
BC007858.2 | - | 3 | 1179 | 4299832 | ||
BX648811.1 | - | 2 | 1950 | |||
J03634.1 | - | 2 | 1047 | |||
M13436.1 | - | 2 | 1198 | |||
X72498.1 | - | 1 | 32 |