Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009272.4 | NP_033298.1 | 4 | 501 | ||
GenBank mRNA | AK028359.1 | - | 4 | 512 | ||
AK051101.1 | - | 4 | 497 | |||
AK146395.1 | - | 4 | 458 | |||
BC005566.1 | - | 4 | 505 | 3966710 | ||
L19311.1 | - | 4 | 469 |