Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136081.2 | NP_001129553.1 | 10 | 1455 | ||
NM_182990.4 | NP_892035.2 | 10 | 1584 | |||
GenBank mRNA | AK146446.1 | - | 10 | 1507 | ||
AK146585.1 | - | 10 | 1504 | |||
AK146607.1 | - | 10 | 1487 | |||
AK146691.1 | - | 10 | 1486 | |||
AK147005.1 | - | 10 | 1515 | |||
AK158552.1 | - | 10 | 1455 | |||
S50213.1 | - | 9 | 1329 |