Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009281.3 | NP_033307.2 | 13, 14 | 1601 | ||
GenBank mRNA | AF011758.2 | - | 1550 | |||
AK029078.1 | - | 13, 14 | 1593 | |||
AK029926.1 | - | 13, 14 | 1866 | |||
BC037658.1 | - | 12, 13 | 1511 | 5356405 |