Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011484.2 | NP_035614.1 | 5 | 581 | ||
GenBank mRNA | AK035924.1 | - | 5 | 504 | ||
AK144987.1 | - | 5 | 598 | |||
AK147832.1 | - | 5 | 491 | |||
AK157767.1 | - | 5 | 528 | |||
AK162431.1 | - | 5 | 528 | |||
AK163720.1 | - | 5 | 528 | |||
AK163922.1 | - | 5 | 518 | |||
AK169170.1 | - | 5 | 588 | |||
BC044666.1 | - | 5 | 581 | 4950569 | ||
BC055326.1 | - | 5 | 527 | 6416552 | ||
U43900.1 | - | 5 | 438 |