Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128928.1 | NP_001122400.1 | 5, 6 | 1174 | ||
NM_002194.3 | NP_002185.1 | 4, 5 | 1030 | |||
XM_005246532.1 | XP_005246589.1 | 4, 5 | 811 | |||
GenBank mRNA | AB451298.1 | - | 3, 4 | 474 | ||
AK093560.1 | - | 6, 7 | 1132 | |||
BC015496.1 | - | 3, 4 | 540 | 3845956 | ||
BC106006.1 | - | 4, 5 | 574 | 4472604 | ||
CR456840.1 | - | 3, 4 | 474 | |||
CR541707.1 | - | 3, 4 | 474 | |||
KJ535023.1 | - | 3, 4 | 474 | |||
KJ535042.1 | - | 3, 4 | 473 | |||
KU178146.1 | - | 3, 4 | 474 | |||
L08488.1 | - | 4, 5 | 800 |