Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009290.2 | NP_033316.1 | 1, 2 | 107 | ||
GenBank mRNA | BC119212.1 | - | 1, 2 | 107 | 8733961 | |
BC120517.1 | - | 1, 2 | 107 | 40129440 | ||
Z68889.1 | - | 1, 2 | 122 |