Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suclg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011507.3 | NP_035637.2 | 4 | 439 | ||
GenBank mRNA | AF058956.1 | - | 3 | 281 | ||
AK042987.1 | - | 4 | 394 | |||
AK045176.1 | - | 4 | 392 | |||
AK156263.1 | - | 4 | 394 | |||
AK167136.1 | - | 4 | 394 | |||
AK167339.1 | - | 4 | 678 | |||
AK172561.1 | - | 4 | 394 | |||
AK172633.1 | - | 4 | 394 | |||
BC043312.1 | - | 2 | 134 | |||
BC054425.1 | - | 3 | 192 | |||
BC080781.1 | - | 4 | 394 | 30470296 | ||
BI985474.1 | - | 4 | 431 | |||
EU234009.1 | - | 4 | 346 |