Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Supt4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009296.1 | NP_033322.1 | 5 | 340 | ||
GenBank mRNA | AK002990.1 | - | 5 | 442 | ||
AK146235.1 | - | 6 | 497 | |||
AK167941.1 | - | 5 | 354 | |||
AK168229.1 | - | 6 | 497 | |||
BC024391.1 | - | 5 | 335 | 5011064 | ||
BC057949.1 | - | 312 | ||||
BC061174.1 | - | 5 | 341 | 6592996 | ||
BC087923.1 | - | 5 | 316 | 30308013 | ||
BC141092.1 | - | 5 | 378 | 9055704 | ||
U43154.1 | - | 5 | 305 |
Gene Symbol: Gm3258 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011509.2 | NP_035639.1 | 1 | 575 |