Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm3258 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011509.2 | NP_035639.1 | 1 | 349 |
Gene Symbol: Supt4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC057949.1 | - | 86 |