Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Surf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013678.2 | NP_038706.1 | 4 | 373 | ||
GenBank mRNA | AK004931.1 | - | 1 | 2186 | ||
AK157125.1 | - | 4 | 387 | |||
AK165559.1 | - | 4 | 380 | |||
AK166181.1 | - | 4 | 402 | |||
AK172018.1 | - | 4 | 376 | |||
AK177886.1 | - | 50 | ||||
AK188020.1 | - | 2 | 50 | |||
BC049779.1 | - | 3 | 1704 | |||
BC096401.1 | - | 4 | 373 | 1365532 |