Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009317.3 | NP_033343.1 | 2 | 135 | ||
GenBank mRNA | AK018448.1 | - | 2 | 136 | ||
BC110625.2 | - | 2 | 105 | 40051579 |