Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tbx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011536.2 | NP_035666.2 | 7 | 926 | ||
NM_172798.1 | NP_766386.1 | 6 | 876 | |||
GenBank mRNA | AB221665.1 | - | 7 | 926 | ||
AK028407.1 | - | 6 | 810 | |||
AK139949.1 | - | 6 | 849 | |||
AK147984.1 | - | 6 | 794 | |||
AY075134.1 | - | 7 | 926 |