Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009328.2 | NP_033354.2 | 1 | 332 | ||
GenBank mRNA | BC027533.1 | - | 1 | 332 | 5143248 | |
U18658.1 | - | 1 | 369 |