Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfap2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025305.2 | NP_001020476.1 | 4 | 1000 | ||
NM_001286340.1 | NP_001273269.1 | 4 | 965 | |||
NM_009334.3 | NP_033360.2 | 4 | 965 | |||
GenBank mRNA | AK017373.1 | - | 4 | 967 | ||
AK028906.1 | - | 4 | 920 | |||
AK032045.1 | - | 4 | 1002 | |||
X78197.1 | - | 4 | 891 |