Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ubtf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK004961.1 | - | 15, 16 | 1792 | ||
AK038139.1 | - | 11, 12 | 2922 | |||
AK128947.1 | - | 15, 16 | 1819 | |||
AK158861.1 | - | 15, 16 | 1791 | |||
AK158935.1 | - | 16, 17 | 1707 | |||
BQ572941.1 | - | 6, 7 | 663 | |||
CA977198.1 | - | 1, 2 | 113 | |||
X60831.1 | - | 15, 16 | 1770 |