Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcp10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011553.4 | NP_035683.2 | 9 | 1175 | ||
GenBank mRNA | AK161455.1 | - | 9 | 1146 | ||
BC052353.1 | - | 9 | 1290 | 6773668 |