Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291765.1 | NP_001278694.1 | 3, 4 | 201 | ||
NM_009361.3 | NP_033387.1 | 3, 4 | 262 | |||
GenBank mRNA | AK019997.1 | - | 3, 4 | 202 | ||
AK030820.1 | - | 3, 4 | 263 | |||
BC058633.1 | - | 3, 4 | 245 | |||
BC132567.1 | - | 2, 3 | 107 | 40130844 | ||
BC132569.1 | - | 2, 3 | 107 | 40130846 | ||
BC145344.1 | - | 2, 3 | 107 | 9053900 | ||
X72310.1 | - | 2, 3 | 136 |