Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009367.3 | NP_033393.2 | 6, 7 | 2308 | ||
GenBank mRNA | AK029306.1 | - | 4, 5 | 520 | ||
AK141552.1 | - | 7, 8 | 1691 | |||
AK159690.1 | - | 6, 7 | 872 | |||
BC011055.1 | - | 4, 5 | 671 | 3585774 | ||
BC011170.1 | - | 6, 7 | 1216 | 4035366 | ||
X57413.1 | - | 6, 7 | 2307 |