Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tmpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080129.2 | NP_001073598.1 | 3 | 833 | ||
NM_001080130.2 | NP_001073599.1 | 3 | 833 | |||
NM_001080131.2 | NP_001073600.1 | 3 | 833 | |||
NM_001080132.2 | NP_001073601.1 | 3 | 833 | |||
NM_001283048.1 | NP_001269977.1 | 3 | 516 | |||
NM_011605.3 | NP_035735.2 | 3 | 833 | |||
GenBank mRNA | AA041637.1 | - | 1 | 60 | ||
AK011988.1 | - | 3 | 326 | |||
AK011992.1 | - | 3 | 326 | |||
AK141845.1 | - | 3 | 783 | |||
AK144642.1 | - | 3 | 793 | |||
AK146903.1 | - | 3 | 800 | |||
AK150305.1 | - | 3 | 781 | |||
AK150515.1 | - | 3 | 768 | |||
AK151936.1 | - | 3 | 785 | |||
AK152737.1 | - | 3 | 785 | |||
AK153281.1 | - | 3 | 768 | |||
AK156206.1 | - | 3 | 783 | |||
AK159161.1 | - | 3 | 429 | |||
AK165282.1 | - | 3 | 783 | |||
AK165851.1 | - | 3 | 790 | |||
AK192607.1 | - | 3 | 272 | |||
BC141062.1 | - | 3 | 551 | 9055674 | ||
BE852115.1 | - | 3 | 274 | |||
CN668856.1 | - | 1 | 36 | |||
U39073.1 | - | 3 | 736 | |||
U39074.1 | - | 3 | 764 | |||
U39075.1 | - | 3 | 764 | |||
U39076.1 | - | 3 | 764 | |||
U39077.1 | - | 3 | 764 | |||
U39078.1 | - | 3 | 747 |