Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011608.1 | NP_035738.1 | 3 | 632 | ||
GenBank mRNA | AF061505.1 | - | 3 | 632 | ||
AK020247.1 | - | 3 | 630 | |||
BC027519.1 | - | 3 | 503 | 1349977 |