Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002202.2 | NP_002193.2 | 6 | 1556 | ||
XM_011543380.2 | XP_011541682.1 | 5 | 2674 | |||
GenBank mRNA | AK290742.1 | - | 6 | 1274 | ||
BC031213.1 | - | 6 | 1557 | 5269369 | ||
S70721.1 | - | 6 | 1013 | |||
U07559.1 | - | 6 | 1247 |