Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289895.1 | NP_001276824.1 | 23, 24 | 3672 | ||
NM_027884.3 | NP_082160.3 | 25, 26 | 3733 | |||
GenBank mRNA | AK004758.1 | - | 2, 3 | 260 | ||
BC052740.1 | - | 4, 5 | 498 | |||
BC055076.1 | - | 2, 3 | 310 |