Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tpi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009415.2 | NP_033441.2 | 7 | 883 | ||
GenBank mRNA | AK002270.1 | - | 7 | 745 | ||
AK008034.1 | - | 3 | 243 | |||
AK008373.1 | - | 3 | 240 | |||
AK010808.1 | - | 7 | 753 | |||
AK146013.1 | - | 7 | 742 | |||
AK149768.1 | - | 7 | 747 | |||
AK150735.1 | - | 7 | 744 | |||
AK159741.1 | - | 7 | 747 | |||
AK167437.1 | - | 7 | 748 | |||
AK168446.1 | - | 7 | 747 | |||
AK168756.1 | - | 7 | 746 | |||
AK199996.1 | - | 1 | 32 | |||
BC004024.1 | - | 7 | 731 | |||
BC046761.1 | - | 7 | 734 | 5693696 | ||
X53333.1 | - | 7 | 745 |