Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009419.3 | NP_033445.2 | 3 | 748 | ||
GenBank mRNA | AF049890.1 | - | 2 | 694 | ||
AK036436.1 | - | 2 | 677 | |||
AK075672.1 | - | 3 | 748 | |||
AK149379.1 | - | 3 | 752 | |||
AK149502.1 | - | 3 | 711 | |||
AK150082.1 | - | 3 | 708 | |||
AK163461.1 | - | 3 | 712 | |||
BC003721.1 | - | 3 | 739 | 3494892 |