Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011629.3 | NP_035759.3 | 5 | 619 | ||
GenBank mRNA | AK146891.1 | - | 5 | 632 | ||
AK149374.1 | - | 5 | 534 | |||
BC090662.1 | - | 5 | 595 | 30682075 | ||
BC094580.1 | - | 5 | 537 | 30602939 | ||
L26957.1 | - | 4 | 428 | |||
U30482.1 | - | 4 | 428 | |||
Y11436.1 | - | 4 | 1067 |