Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tnfsf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009425.2 | NP_033451.1 | 2 | 334 | ||
GenBank mRNA | AK142960.1 | - | 2 | 336 | ||
AK144931.1 | - | 351 | ||||
AK153581.1 | - | 2 | 384 | |||
AK157633.1 | - | 335 | ||||
BC011469.1 | - | 2 | 300 | |||
BC138807.1 | - | 2 | 308 | 8861829 | ||
BC138808.1 | - | 2 | 308 | 8861830 | ||
BY734631.1 | - | 2 | 351 | |||
U37522.1 | - | 2 | 310 |