Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trap1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011635.1 | NP_035765.1 | 2 | 815 | ||
GenBank mRNA | AK146737.1 | - | 2 | 630 | ||
BC051644.1 | - | 2 | 608 | 30022193 | ||
M36387.1 | - | 2 | 815 |