Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126330.1 | NP_001119802.1 | 6 | 1047 | ||
NM_001126331.1 | NP_001119803.1 | 6 | 1047 | |||
NM_011642.3 | NP_035772.2 | 8 | 1337 | |||
GenBank mRNA | AB221594.1 | - | 7 | 929 | ||
AK014503.1 | - | 1 | 26 | |||
AK017412.1 | - | 5 | 578 | |||
AK139633.1 | - | 6 | 1048 | |||
BC066045.1 | - | 6 | 934 | 6812399 | ||
BC069182.1 | - | 2 | 194 | |||
Y19234.1 | - | 8 | 1338 | |||
Y19235.1 | - | 6 | 1041 |