Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002205.4 | NP_002196.4 | 17 | 1827 | ||
GenBank mRNA | AK291570.1 | - | 17 | 1772 | ||
AK312414.1 | - | 17 | 1827 | |||
BC008786.2 | - | 17 | 1812 | 3629647 | ||
BX647780.1 | - | 7 | 1270 | |||
M13918.2 | - | 17 | 1714 | |||
X06256.1 | - | 17 | 1782 |